Learn More
USA Scientific Inc Large block for 50 mL tubes
Catalog No. 502745506
Encompass
Large aluminum block for 50 mL tubes. Holds four 50 mL tubes.
Catalog No. 50-274-5506 Supplier USA Scientific Inc Supplier No. 91247050
Large aluminum block for 50 mL tubes. Holds four 50 mL tubes.