Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Large Blocking Arm for Valve Lockout

Catalog No. 19129183 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-183 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-183 Supplier Brady™ Supplier No. 65403
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made of industrial-grade steel and nylon

  • Versatile, lightweight and rugged Blocking Arm is easy to use, and can lock out numerous types of valves
  • Made of rugged industrial-grade steel and nylon for extra impact and chemical resistance
  • Specifications

    Product Type Valve Lockouts
    Color Red
    Material Steel
    Dimensions (D x W x H) 37 x 8.82 x 7.05 in.
    For Use With (Application) Lockout-Tagout Stations and Kits
    Quantity 1 Ea.
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

    Adhere to
    Safety Regulations and Best Practices

    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.