Learn More
Andwin Scientific Large Protection ESD Lab Coat
SDP
Catalog No. NC1346180
Encompass_Preferred
VWR ESD blue is made of 98% polyester-2% carbon with 3 pockets, knit cuffs, snap closure. 44in long, Large
This item is not returnable. View return policy
Catalog No. NC1346180 Supplier Andwin Scientific Supplier No. 10769340
VWR ESD blue is made of 98% polyester-2% carbon with 3 pockets, knit cuffs, snap closure. 44in long, Large