Learn More
Mitegen Large Blunt Tweezers, 12-Inch Length, Stainless Steel with Serrated Tips, For Secure Handling in Laboratory & Cryogenic Environments
SDP
Catalog No. NC2675832
Encompass
These Large Blunt Tweezers, measuring a substantial 12 inches in length, are robust instruments designed for secure and precise handling of various items in demanding laboratory and industrial settings. Constructed from high-quality stainless steel, they offer excellent durability, corrosion resistance, and are easy to clean, making them suitable for use in diverse environments, including cryogenic applications. The blunt, serrated tips provide a firm, non-slip grip on larger or irregularly shaped objects, reducing the risk of accidental drops or damage during transfers and manipulations. Their extended length allows for safe handling of materials in deep containers, liquid nitrogen dewars, or in situations requiring increased reach while maintaining a safe distance. These versatile tweezers are an indispensable tool for a wide range of tasks where a strong, secure, and controlled grip is paramount.
Catalog No. NC2675832 Supplier Mitegen Supplier No. MCEMTW12LB
These Large Blunt Tweezers, measuring a substantial 12 inches in length, are robust instruments designed for secure and precise handling of various items in demanding laboratory and industrial settings. Constructed from high-quality stainless steel, they offer excellent durability, corrosion resistance, and are easy to clean, making them suitable for use in diverse environments, including cryogenic applications. The blunt, serrated tips provide a firm, non-slip grip on larger or irregularly shaped objects, reducing the risk of accidental drops or damage during transfers and manipulations. Their extended length allows for safe handling of materials in deep containers, liquid nitrogen dewars, or in situations requiring increased reach while maintaining a safe distance. These versatile tweezers are an indispensable tool for a wide range of tasks where a strong, secure, and controlled grip is paramount.