Learn More
Grainger Large Bore Core Remover
Catalog No. 502753149
Encompass_Preferred
The Haltec Large Bore Core Remover is a tool used to remove large bore cores from engines or other machinery. It has a capacity of removing cores up to 8 inches in diameter. The tool is made from drop-forged alloy steel for durability. It features a T-handle for applying torque and has a hex-shaped shank to fit a wrench if needed. The core remover has a threaded end to screw into the core and extract it. This heavy-duty tool allows for easy core removal from large bore engines or machinery.
Catalog No. 50-275-3149 Supplier Grainger Supplier No. 141A53
The Haltec Large Bore Core Remover is a tool used to remove large bore cores from engines or other machinery. It has a capacity of removing cores up to 8 inches in diameter. The tool is made from drop-forged alloy steel for durability. It features a T-handle for applying torque and has a hex-shaped shank to fit a wrench if needed. The core remover has a threaded end to screw into the core and extract it. This heavy-duty tool allows for easy core removal from large bore engines or machinery.