Learn More
Grainger Large Bore Core Remover
Catalog No. 502753008
Encompass_Preferred
The Uni-Weld Valve Core Removal Tool is a handheld tool used to remove valve cores from tires and inflatable objects. It features a knurled body for grip and a slotted end to fit valve cores. The tool is made from heat treated steel and removes valve cores without damage. It works on all standard valve cores including rubber snap-in, metal clamp-in, and threaded screw-in types. The compact 4 inch size fits easily in a pocket or toolbox for portability. This durable American made tool allows quick and easy valve core removal for tire repairs or maintenance.
Catalog No. 50-275-3008 Supplier Grainger Supplier No. 141C72
The Uni-Weld Valve Core Removal Tool is a handheld tool used to remove valve cores from tires and inflatable objects. It features a knurled body for grip and a slotted end to fit valve cores. The tool is made from heat treated steel and removes valve cores without damage. It works on all standard valve cores including rubber snap-in, metal clamp-in, and threaded screw-in types. The compact 4 inch size fits easily in a pocket or toolbox for portability. This durable American made tool allows quick and easy valve core removal for tire repairs or maintenance.