Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Large Bore Female Luer with 1/4 in. ID - Natural Kynar - QC
SDP

Catalog No. 01000178 Shop All Fisherbrand Products
Catalog No. 01-000-178

missing translation for 'validMainframeMsg'

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Large bore luers offer a higher flow rates than standard luers. 

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • RoHS Compliant
  • Gamma Stable
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Natural Kynar
For Use With (Application) Medical, Pharmaceutical, Industrial applications
Packaging Quantity 100 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Large Bore Luer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.