Learn More
Daigger & Company Large Capacity Round Bottle, 2 L, 6/cs
SDP
Catalog No. NC1869588
Encompass
Wide-Mouth High-Density Polyethylene Bottles are excellent for dry chemicals or water sampling. Wide-Mouth Polypropylene Bottles are specifically designed for applications requiring autoclaving of large volumes. Note: bottles are not leakproof.
Catalog No. NC1869588 Supplier Daigger & Company Supplier No. NAL21200005
Wide-Mouth High-Density Polyethylene Bottles are excellent for dry chemicals or water sampling. Wide-Mouth Polypropylene Bottles are specifically designed for applications requiring autoclaving of large volumes. Note: bottles are not leakproof.