Learn More
JDM LAB SOLUTIONS LLC LARGE CAPACITY ORBITAL SHAKER
Catalog No. NC2307317
Encompass
Large Capacity Orbital Shaker SYC-2102AFeatures Ergonomic design with speed control and timer Automatically recovers from power outages Wide range of accessories available Digital control and LED/LCD display Designed and built for continuous use Gentle speed up and slow down Safe for use inside standard incubators No vibration or heat output Built for heavy workloadsOrbital Shakers may be used for several applications including culturing microbes, washing blots, and general mixing. Our Shakers are designed and built for a heavy workload and provides continuous shaking with no vibration or heat output. It can be placed into a standard incubator, converting the incubator into a shaking incubator.Speed: 30 - 300 RPMPlatform Dimensions: 15.7 x 15.7 inches
Catalog No. NC2307317 Supplier JDM LAB SOLUTIONS LLC Supplier No. SYC2102A
Large Capacity Orbital Shaker SYC-2102AFeatures Ergonomic design with speed control and timer Automatically recovers from power outages Wide range of accessories available Digital control and LED/LCD display Designed and built for continuous use Gentle speed up and slow down Safe for use inside standard incubators No vibration or heat output Built for heavy workloadsOrbital Shakers may be used for several applications including culturing microbes, washing blots, and general mixing. Our Shakers are designed and built for a heavy workload and provides continuous shaking with no vibration or heat output. It can be placed into a standard incubator, converting the incubator into a shaking incubator.Speed: 30 - 300 RPMPlatform Dimensions: 15.7 x 15.7 inches