Learn More
Research Products International Corp Large Capacity Sample Blocks, 45 x 0.5ml and 32 x 1.5ml Tubes
SDP
Catalog No. 501977068
Encompass
Ideal dry bath incubator can freeze, chill or heat a variety of tube sizes including 0.2, 0.5, 2.0, 15 and 50ml. Aluminum blocks are also available for assay plates, test tubes and round bottom flasks. Compact and portable design requires minimal bench space.
Blocks are not included with unit. Order separately.
Catalog No. 50-197-7068 Supplier Research Products International Corp Supplier No. 348273
Ideal dry bath incubator can freeze, chill or heat a variety of tube sizes including 0.2, 0.5, 2.0, 15 and 50ml. Aluminum blocks are also available for assay plates, test tubes and round bottom flasks. Compact and portable design requires minimal bench space.
Blocks are not included with unit. Order separately.