Learn More
Research Products International Corp Large Capacity Sample Blocks, 96 x 0.2ml Micro-Tubes and Plates
SDP
Catalog No. 501977128
Encompass
Ideal dry bath incubator can freeze, chill or heat a variety of tube sizes including 0.2, 0.5, 2.0, 15 and 50ml. Aluminum blocks are also available for assay plates, test tubes and round bottom flasks. Compact and portable design requires minimal bench space.
Blocks are not included with unit. Order separately.
Catalog No. 50-197-7128 Supplier Research Products International Corp Supplier No. 348274
Ideal dry bath incubator can freeze, chill or heat a variety of tube sizes including 0.2, 0.5, 2.0, 15 and 50ml. Aluminum blocks are also available for assay plates, test tubes and round bottom flasks. Compact and portable design requires minimal bench space.
Blocks are not included with unit. Order separately.