Learn More
Cole-Parmer LARGE-CAPACITY STIRRER, 120 VA
Catalog No. NC2046911 Shop All Cole-Parmer Products
Encompass
Corning 6795-611 Economical Large-Capacity Stirrer, 120 VAC, 50/60 Hz. https://www.coleparmer.com/i/corning-6795-611-economical-large-capacity-stirrer-120-vac-50-60-hz/8430230#:~:text=Stirs%20up%20to%2038%20liters%20(10%20gallons)%20of%20aqueous%20solution
Catalog No. NC2046911 Supplier Cole-Parmer Supplier No. 8430230
Corning 6795-611 Economical Large-Capacity Stirrer, 120 VAC, 50/60 Hz. https://www.coleparmer.com/i/corning-6795-611-economical-large-capacity-stirrer-120-vac-50-60-hz/8430230#:~:text=Stirs%20up%20to%2038%20liters%20(10%20gallons)%20of%20aqueous%20solution