Learn More
Scientific Plastics LARGE CARBOY CONTAINMENT
SDP
Catalog No. NC1059858
Encompass_Preferred
22” for most countertops.10” trough to fill flasks. 4.9 gal/18L containment.100lb capacity
Catalog No. NC1059858 Supplier Scientific Plastics Supplier No. CB222012001
22” for most countertops.10” trough to fill flasks. 4.9 gal/18L containment.100lb capacity