Learn More
Tewes Corporation 51 x 49 x 73 1.5 Mil Covers
SDP
Catalog No. NC1841713
Encompass_Preferred
Gusset bags are ideal for covering equipment or machinery to keep dust free and dry. They can also be used as box liners or for packaging bulky items. The gusseted sides expand to give extra space for storage and shipping. Bags are perforated on rolls for easy tear off and dispensing and meet FDA and USDA specifications. 75/ROLL.
Catalog No. NC1841713 Supplier Tewes Corporation Supplier No. G10073POR
Gusset bags are ideal for covering equipment or machinery to keep dust free and dry. They can also be used as box liners or for packaging bulky items. The gusseted sides expand to give extra space for storage and shipping. Bags are perforated on rolls for easy tear off and dispensing and meet FDA and USDA specifications. 75/ROLL.