Learn More
Memmert Memmert Climate Chamber HPP 1060eco, 100-240volt Temp Range 0C-70C Humidity Range 10-90%RH D8-40mm silicone entry port with stopper rear wall
SDP
Catalog No. 5000451068
Encompass_Preferred
Memmert Climate Chamber HPP 1060eco, 100-240volt Temp Range 0C-70C Humidity Range 10-90%RH D8-40mm silicone entry port with stopper rear wall
Catalog No. 50-004-51068 Supplier Memmert Supplier No. HPP 1060ECO 100240VOLT
Memmert Climate Chamber HPP 1060eco, 100-240volt Temp Range 0C-70C Humidity Range 10-90%RH D8-40mm silicone entry port with stopper rear wall