Learn More
Memmert Memmert Climate Chamber HPP 1400eco, 100-240Volt, +15C (10C below ambient to +60C, 10-80%RH D8-40mm silicone entry port with stopper rear wall
SDP
Catalog No. 5000451103
Encompass_Preferred
Memmert Climate Chamber HPP 1400eco, 100-240Volt, +15C (10C below ambient to +60C, 10-80%RH D8-40mm silicone entry port with stopper rear wall
Catalog No. 50-004-51103 Supplier Memmert Supplier No. HPP 1400ECO 100240VOLT
Memmert Climate Chamber HPP 1400eco, 100-240Volt, +15C (10C below ambient to +60C, 10-80%RH D8-40mm silicone entry port with stopper rear wall