Learn More
Memmert Memmert Climate Chamber HPP 2200eco, 230 Volt 50/60 Hz, 15 - 60C temp range, 15-80%RH 40mm silicone entry port with stopper rear wall
SDP
Catalog No. 5000451135
Encompass_Preferred
Memmert Climate Chamber HPP 2200eco, 230 Volt 50/60 Hz, 15 - 60C temp range, 15-80%RH 40mm silicone entry port with stopper rear wall
Catalog No. 50-004-51135 Supplier Memmert Supplier No. HPP 2200ECO 230VOLT
Memmert Climate Chamber HPP 2200eco, 230 Volt 50/60 Hz, 15 - 60C temp range, 15-80%RH 40mm silicone entry port with stopper rear wall