Learn More
Electron Microscopy Sciences Large Dissecting Kit
SDP
Catalog No. 5032232
Encompass_Preferred
Dissecting kit includes 12 scalpel blades #11 with #3 Handle, 6 1⁄2" kelly hemostat, straight, 6 1⁄4" kelly hemostat, straight, 4 1⁄2" iris scissors, 51⁄2" dissecting scissors, set of dissecting needle, one straight and one bent, 4 1⁄2" straight fine point serrated jaws forceps, 5 1⁄2" straight blunt tip serrated jaws forceps, 2 mm diameter stainless steel punch, suture hook, and large Wood Handle Spatula EMS # Description Qty/Cs.
Catalog No. 50-322-32 Supplier Electron Microscopy Sciences Supplier No. 7295115
Dissecting kit includes 12 scalpel blades #11 with #3 Handle, 6 1⁄2" kelly hemostat, straight, 6 1⁄4" kelly hemostat, straight, 4 1⁄2" iris scissors, 51⁄2" dissecting scissors, set of dissecting needle, one straight and one bent, 4 1⁄2" straight fine point serrated jaws forceps, 5 1⁄2" straight blunt tip serrated jaws forceps, 2 mm diameter stainless steel punch, suture hook, and large Wood Handle Spatula EMS # Description Qty/Cs.