Learn More
Mopec Large Dissection Toolkit
SDP
Catalog No. 22038245 Shop All Mopec Products
Catalog No. 22-038-245 Supplier Mopec Supplier No. BU003
Add to Cart
Add to Cart
Designed for Pathologists’ Assistants primarily performing dissections on large specimens, the large dissection toolkit includes the essentials for safe grossing with a number 60 scalpel blade.
The Toolkit includes:
  • (2) AJ112: Scalpel Blades, Sterile #60, 100 pack
  • (2) AJ160: BladeFlask EVO Universal Blade Remover
  • (2) BE094: Formaway Formalin. Neutralizing Pads, 11 x 15 in., 25 pack
We recommend ordering this toolkit monthly.