Learn More
Bellco Glass, Inc. Large Flange Drive Adapter Omni Vessels
SDP
Catalog No. 501946196
Encompass_Preferred
316 L Stainless Steel Large Flange Drive Adapter for use with Omni Vessels. For attachment of Overhead Drive Motors. Heavy duty cap assembly for large flange vessels Adapt overhead drives to teflon or sst folding impeller assembly 316 sst design with polyester stainless steel bearings. MADE IN THE USA
Catalog No. 50-194-6196 Supplier Bellco Glass, Inc. Supplier No. 776471000
316 L Stainless Steel Large Flange Drive Adapter for use with Omni Vessels. For attachment of Overhead Drive Motors. Heavy duty cap assembly for large flange vessels Adapt overhead drives to teflon or sst folding impeller assembly 316 sst design with polyester stainless steel bearings. MADE IN THE USA