Learn More
Mitegen Large Foam Dewar, 1400 mL Capacity, Lightweight, Shatterproof, Excellent Thermal Insulation for Cryogenic & Chilling Applications
SDP
Catalog No. 501187301
Encompass
Large Foam Dewar offers a versatile and safe solution for a wide range of laboratory applications requiring temporary cryogenic storage or precise chilling. With a substantial 1400 mL (1.4 Liter) capacity, this dewar is perfectly suited for handling liquid nitrogen, dry ice, ice, or other various cooling agents. Constructed from robust, high-density foam, it provides excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of your sensitive samples or reagents. Its lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops. The dewar features a wide, accessible opening for convenient insertion and retrieval of materials, and its stable base promotes secure placement on any benchtop. Ideal for quick chilling, temporary storage of cryo-vials, and other temperature-sensitive items, this Mitegen foam dewar is a highly reliable and indispensable addition to any scientific setting.
Catalog No. 50-118-7301 Supplier Mitegen Supplier No. MFD1400
Large Foam Dewar offers a versatile and safe solution for a wide range of laboratory applications requiring temporary cryogenic storage or precise chilling. With a substantial 1400 mL (1.4 Liter) capacity, this dewar is perfectly suited for handling liquid nitrogen, dry ice, ice, or other various cooling agents. Constructed from robust, high-density foam, it provides excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of your sensitive samples or reagents. Its lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops. The dewar features a wide, accessible opening for convenient insertion and retrieval of materials, and its stable base promotes secure placement on any benchtop. Ideal for quick chilling, temporary storage of cryo-vials, and other temperature-sensitive items, this Mitegen foam dewar is a highly reliable and indispensable addition to any scientific setting.