Learn More
World Precision Instrument Large FORT Force Transducer-FORT1000
SDP
Catalog No. 502538800
Encompass
This rigid-lever force transducer transforms applied force into proportional voltage. Using balanced strain gauges, FORT transducers produce linear output voltage vs. applied force input with very little deflection. To use, clamp the handle of the FORT transducer in a horizontal position and apply the forces to be measured to a rivet or hook mounted in the hole at the end of the flat sensing leaf.
The transducer is highly resistant to mechanical damage. Its body is machined from aluminum and mounts to a standard ring stand or, with the handle removed, clamp mounts.
Catalog No. 50-253-8800 Supplier World Precision Instrument Supplier No. FORT1000
This rigid-lever force transducer transforms applied force into proportional voltage. Using balanced strain gauges, FORT transducers produce linear output voltage vs. applied force input with very little deflection. To use, clamp the handle of the FORT transducer in a horizontal position and apply the forces to be measured to a rivet or hook mounted in the hole at the end of the flat sensing leaf.
The transducer is highly resistant to mechanical damage. Its body is machined from aluminum and mounts to a standard ring stand or, with the handle removed, clamp mounts.