Learn More
Ampco Large Gasket Knife with Looped End
SDP
Catalog No. 19174139 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
49 in.
Catalog No. Length (English)
19-174-139 49 in.
1 options
Catalog No. 19-174-139 Supplier Ampco Supplier No. K54
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant gasket knife.
  • Please see image for additional information regarding measurements.
  • Tip width (a): 1 and 1/2 in.
  • Tip length (b): 31 and 1/2 in.
  • Overall length: 49 in.
Specifications
Product Type Gasket Knife
Length (English) Blade 31 -1/2 in.
Length (English) 49 in.
Quantity 1 Ea.