Learn More
Thomas Scientific Large Gel Tray Set (12.5 x 12cm)
SDP
Catalog No. 501459309
Encompass
Multichannel Performance Labnet's ENDURO™ Gel XL includes all components necessary to cast and run horizontal gels: gel trays, combs, casting stand, gel tank and digital power supply. The migration tank, gel trays and multichannel combs are molded for leak-proof performance and a long service life. The direct connection of the power supply into the gel box eliminates the need for power leads. Power - 150v/400ma Control - set voltage, amperage and time Flexibility - runs 6x6cm to 12.5x12cm gels Throughput - up to 112 samples at once Convenience - casting set and combs included Pressing the eject button separates the gel box and power supply. Reconnecting is easy-just press the two pieces together. The power supply can be set to either constant voltage or constant current with automatic crossover between the two parameters. Parameters are shown on the large digital display that is easily visible from a distance. There are three different gel options for the ENDURO™ Gel XL
Catalog No. 50-145-9309 Supplier Thomas Scientific Supplier No. 1196M13
Multichannel Performance Labnet's ENDURO™ Gel XL includes all components necessary to cast and run horizontal gels: gel trays, combs, casting stand, gel tank and digital power supply. The migration tank, gel trays and multichannel combs are molded for leak-proof performance and a long service life. The direct connection of the power supply into the gel box eliminates the need for power leads. Power - 150v/400ma Control - set voltage, amperage and time Flexibility - runs 6x6cm to 12.5x12cm gels Throughput - up to 112 samples at once Convenience - casting set and combs included Pressing the eject button separates the gel box and power supply. Reconnecting is easy-just press the two pieces together. The power supply can be set to either constant voltage or constant current with automatic crossover between the two parameters. Parameters are shown on the large digital display that is easily visible from a distance. There are three different gel options for the ENDURO™ Gel XL