Learn More
Ted Pella Inc Large Glass Slides 4 x 3" (102 x 76mm), 50/pk
SDP
Catalog No. NC1824406
Encompass
These large microscope slides are specially made for larger tissue sections, such as whole brain sections, to enable uninterrupted examination w/o loss of tissue. Standard thickness is 1.2mm, cut ends (not available with frosted ends). Made from optical grade soda lime glass. Sizes available are 4 x 3" up to 7 x 5".
Catalog No. NC1824406 Supplier Ted Pella Inc Supplier No. 26023050
These large microscope slides are specially made for larger tissue sections, such as whole brain sections, to enable uninterrupted examination w/o loss of tissue. Standard thickness is 1.2mm, cut ends (not available with frosted ends). Made from optical grade soda lime glass. Sizes available are 4 x 3" up to 7 x 5".