Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somni Scientific™ Large Induction Chamber, Clear, Passive
SDP

Catalog No. 04777232 Shop All Somni Scientific Products
Change view
Click to view available options
Animal:
Mouse, Rat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
04-777-232 Mouse, Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-232 Supplier Somni Scientific™ Supplier No. IC3012

missing translation for 'validMainframeMsg'

The Large Induction Chamber, Clear is available in Active or Passive styles. Constructed of durable, shatter resistant PET-G and supplied with color coded inlet and outlet ports. The slide top lid keeps anesthetic gases from being drawn into the workplace. Convenient one-handed operation.

  • Active style prevents unnecessary exposure when used with active waste gas removal system
  • Slide top lid keeps anesthetic from being “drawn” into workplace
  • One-handed operation
  • Color coded inlet and outlet ports
  • Compatible with 19 mm tubing and 15 mm adapters
  • Constructed of shatter resistant Pet-G
  • Can be cleaned with common clinical disinfectants

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
Animal Mouse, Rat
Type Induction Chamber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.