Learn More
World Precision Instrument Large Knob Cover Silicone
SDP
Catalog No. 502539228
Encompass
Silicone handle covers for use with the SurgiScope Surgical Microscope. The silicone handle covers protect the handles and knobs of your microscope during surgery and can be easily removed for cleaning.
Catalog No. 50-253-9228 Supplier World Precision Instrument Supplier No. 505130
Silicone handle covers for use with the SurgiScope Surgical Microscope. The silicone handle covers protect the handles and knobs of your microscope during surgery and can be easily removed for cleaning.