Learn More
Cole-Parmer Flame-Resistant Lab Coat, Light Blue, large
Supplier:  Cole-Parmer 8646832
Encompass_Preferred
VF Workwear KEL2LB-LARGE Flame-Resistant Lab Coat, Light Blue, large, Flame-resistant lab coats protect you when working with flammable chemicals, Constructed of 100% cotton, lab coats feature three outside pockets, side vent openings, hemmed sleeve ends, and a lapel collar. Rated to Arc Thermal Protective Value (ATPV) of 7.7 calories/cm2 and a Hazard Risk Category (HRC) of HRC1.
Catalog No. NC1379353