Learn More
Electronic Imaging Materials Inc 667 Vial CryoLabel 1.25 inch x 1.00 inch, White, Roll of 5000, 3 inch core
SDP
Catalog No. NC9911449
Encompass_Preferred
1.25 inch x 1.00 inch CryoLabel designed for vials and plates stored at extremely low temperatures. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer printing and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC9911449 Supplier Electronic Imaging Materials Inc Supplier No. 667125100
1.25 inch x 1.00 inch CryoLabel designed for vials and plates stored at extremely low temperatures. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer printing and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.