Learn More
Mopec Large Magnetic Powder Applicator
SDP
Catalog No. 22446218 Shop All Mopec Products
Change view
Click to view available options
Length (English):
5.53 in.
Catalog No. Length (English)
22-446-218 5.53 in.
1 options
Catalog No. 22-446-218 Supplier Mopec Supplier No. FS020
Add to Cart
Add to Cart
Features a durable, High-density polyethylene shell with a Long-lasting, Spring-loaded steel handle equipped with a powerful neodymium magnet.
  • Spring-loaded handle
  • Plexiglas protective tube
  • Made from High-density polyethylene, neodymium, and Long-lasting steel.
  • Supports hundreds of applications
  • Helps expedite crime scene processing
  • Designed for repeated use and long-lasting performance.
  • Holds more magnetic powder than standard applicators.
Specifications
Product Type Magnetic Powder Applicator
Length (English) 5.53 in.