Learn More
Brady™ Large Marine Spill Truck
Catalog No. 19103129 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
153 gal./Bale
Catalog No. Capacity (English)
19-103-129 153 gal./Bale
1 options
Catalog No. 19-103-129 Supplier Brady™ Supplier No. SKMLT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Prepare yourself for over spray, leaks and drips on water. The Brady™ Large Marine Spill Truck has everything you need and is wheel mounted for maximum mobility.
  • The yellow, polyethylene Marine Large Spill Truck is chemical and water resistant
  • Wheel mounted for maximum mobility
  • Spill trucks provide the highest level of spill readiness
  • Not suitable for outdoor use
  • For use when cleaning up a spill on the water
Specifications
Capacity (English) 153 gal./Bale
Material Polyethylene
Type Large Marine Spill Truck
For Use With (Application) Over spray, leaks, drips and spill response
Includes (400) - 15 in. x 19 in. Pads, (4) - 5 in. x 10 ft. Booms, (1) - 19 in. x 100 ft. Sweep, (6) - Pair Nitrile gloves, (15) - Disposal BagsGogglesEmergency Response Handbook
Mounting Wheel-mounted
Packaging Quantity 1 Kit