Learn More
Electron Microscopy Sciences High Opacity Paint Markers Large Black
SDP
Catalog No. 50949088
Encompass_Preferred
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.
Catalog No. 50-949-088 Supplier Electron Microscopy Sciences Supplier No. 72169B2
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.