Learn More
Electron Microscopy Sciences High Opacity Paint Markers Large Red
SDP
Catalog No. 50949086
Encompass_Preferred
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.
Catalog No. 50-949-086 Supplier Electron Microscopy Sciences Supplier No. 72167R2
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.