Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Large Microtube Rack Holder - Two Column
SDP

Catalog No. 22022597
Change view
Click to view available options
Positions:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
22-022-597 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-597 Supplier Poltex Supplier No. 5LGMICRODBL
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poltex Large Microtube Rack Holder is designed to hold large microtube racks. Made of durable, cleanable PETG construction. Does not include microtube racks.

  • Holds large microtube racks
  • Durable, cleanable PETG construction
  • Microtube racks are not included

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Microtubes
Material PETG
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
Length (Metric) 236 mm
Product Type Microtube Rack Tray
Shelves 4 Tiers
Height (Metric) 273 mm
Width (Metric) 319.28 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.