Learn More
Euthanex The EZ-1785 Sure-Seal Large Mouse/Rat Chamber
Catalog No. NC0955931
Encompass_Preferred
The EZ-1785 Sure-Seal Large Mouse/Rat Chamber is constructed of heavy-duty, clear acrylic plastic with a non-skid bottom and simple snap latch top. It can be used to anestitize animals for pre-surgery or use for euthanasia when the home cage is unavailable. Dimensions: 11.25? L x 7? W x 6? H.
Catalog No. NC0955931 Supplier Euthanex Supplier No. EZ1785
The EZ-1785 Sure-Seal Large Mouse/Rat Chamber is constructed of heavy-duty, clear acrylic plastic with a non-skid bottom and simple snap latch top. It can be used to anestitize animals for pre-surgery or use for euthanasia when the home cage is unavailable. Dimensions: 11.25? L x 7? W x 6? H.