Learn More
Chemglass Life Sciences Funnel, Process Reactor, Large, Polypropylene, 14.1L Capacity, 350mm Top, 80mm Flange
SDP
Catalog No. 501531238
Encompass_Preferred
Funnel, Process Reactor, Large, Polypropylene, 14.1L Capacity, 350mm Top, 80mm Flange. Large funnels have a 80mm flange fused to the bottom of the funnel. This design makes them ideal for filling Chemglass's process reactors by clamping the funnel directly to the powder addition port. The wide mouth funnel has a large opening to optimize flow. The 4.3L size has a handle at the rim. Main body and flange are made from Polypropylene. Funnels are autoclavable at 121C (250F).
Catalog No. 50-153-1238 Supplier Chemglass Life Sciences Supplier No. CG1767R814
Funnel, Process Reactor, Large, Polypropylene, 14.1L Capacity, 350mm Top, 80mm Flange. Large funnels have a 80mm flange fused to the bottom of the funnel. This design makes them ideal for filling Chemglass's process reactors by clamping the funnel directly to the powder addition port. The wide mouth funnel has a large opening to optimize flow. The 4.3L size has a handle at the rim. Main body and flange are made from Polypropylene. Funnels are autoclavable at 121C (250F).