Learn More
World Precision Instrument Large Probe Holder
SDP
Catalog No. 502539164
Encompass
  • Large Probe Holder supports shafts with diameters from 6.5 mm up to 13 mm (0.5 inch).
Catalog No. 50-253-9164 Supplier World Precision Instrument Supplier No. 505251
  • Large Probe Holder supports shafts with diameters from 6.5 mm up to 13 mm (0.5 inch).