Learn More
ZIVIC INSTRUMENTS LARGE RAT BRAIN SLICER MATRIX
Supplier:  ZIVIC INSTRUMENTS BSRLS0021
Encompass
KEY FEATURES:- Slice intervals spaced 2.0mm apart- Use this matrix to section the rat brain into CORONAL slices with thicknesses in increments of 2.0mm- Designed to accommodate for unique anatomical features of the rat brain, including adjacent olfactory/spinal tissue- Include a central sagittal slice channel, allowing the brain to be split into left and right hemispheres- Use with fresh or preserved brain tissue- Autoclavable and designed for long-term use- Matrices of any given type are identical to insure reproducible sections- Comes ready to use, with 10 single edge razor blades provided at no additional cost- Made from high-grade stainless steelUSES INCLUDE:- Dissection / Blocking of the rat brain into coronal sections (or left and right hemispheres)- Sectioning the rat brain prior to or post staining with agents included TTC or X-Gal-stain- Repeated isolation of brain regions for further analysis or slide
Catalog No. NC3376528