Learn More
Electron Microscopy Sciences Vortex Genie Large Sample Head Set
SDP
Catalog No. 50949625
Encompass_Preferred
Snap on these attachments for new ways to use your Vortex-Genie. Large Sample Set (includes: Recessed Platform, Elastic Band, 9-16mm Tube Insert, 14-29mm Tube Insert, 29-37mm Tube Insert, Blank Insert).
Catalog No. 50-949-625 Supplier Electron Microscopy Sciences Supplier No. 71843
Snap on these attachments for new ways to use your Vortex-Genie. Large Sample Set (includes: Recessed Platform, Elastic Band, 9-16mm Tube Insert, 14-29mm Tube Insert, 29-37mm Tube Insert, Blank Insert).