Learn More
OHAUS Large Sample Set Model H302
Catalog No. 01804393 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-393 Vortex Attachment
1 options
Catalog No. 01-804-393 Supplier OHAUS Supplier No. 83041422
Add to Cart
Add to Cart
Sample Set for the Vortex Genie 2 mixers for beakers flasks and large tubes.
  • Includes Recessed Platform 504-0039-00 2.
  • Elastic Bands, an assortment of Foam Inserts.
  • One Blank Foam Insert is included for user customization.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse