Learn More
Synthware™ Large Scale Distillation Appartaus
GREENER_CHOICESDP
Catalog No. 31501058 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
31-501-058 24/40
1 options
Catalog No. 31-501-058 Supplier Synthware™ Supplier No. D502440
Add to Cart
Add to Cart
One-piece design with ith 10/18 thermometer joint and high efficiency cooling jacket.
To be used with 50 mm immersion thermometer with 10/18 joint.
Specifications
For Use With (Application) Organic Synthesis
Type Large Scale
Material Borosilicate Glass
For Use With (Equipment) 25 mm Immersion Thermometer
Quantity 1 Ea.
Standard Taper Joint 24/40
Joint Type Ground Glass
Autoclavable Autoclavable