Learn More
Sklar Instruments Large Lab Type Scalpel Handle, Sklar 4-1/2 in., Stainless Steel. Sklar Large Lab Type Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. EA.
SDP
Catalog No. 501181209 Shop All Sklar Instruments Products
Encompass_Preferred
Large Lab Type Scalpel Handle, Sklar. 4-1/2 in., Stainless Steel. Sklar's Large Lab Type Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle is phenolic versus stainless steel and thicker and shorter than the stainless steel handles.
Catalog No. 50-118-1209 Supplier Sklar Instruments Supplier No. 062925
Large Lab Type Scalpel Handle, Sklar. 4-1/2 in., Stainless Steel. Sklar's Large Lab Type Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle is phenolic versus stainless steel and thicker and shorter than the stainless steel handles.