Learn More
Sklar Instruments Large Lab Type Scalpel Handle, Sklar 5 in., Stainless Steel. Sklar Large Lab Type #4 Scalpel Handle is similar to the #3 Lab Type Scalpel Handle, but compatible with the larger size 20 series disposable blades. EA.
SDP
Catalog No. 501181210 Shop All Sklar Instruments Products
Encompass_Preferred
Large Lab Type Scalpel Handle, Sklar. 5 in., Stainless Steel. Sklar's Large Lab Type #4 Scalpel Handle is similar to the #3 Lab Type Scalpel Handle, but compatible with the larger size 20 series disposable blades.
Catalog No. 50-118-1210 Supplier Sklar Instruments Supplier No. 062930
Large Lab Type Scalpel Handle, Sklar. 5 in., Stainless Steel. Sklar's Large Lab Type #4 Scalpel Handle is similar to the #3 Lab Type Scalpel Handle, but compatible with the larger size 20 series disposable blades.