Learn More
Medicus Health Large Slide Warmer 25” x 8” (63cm x 20cm)
SDP
Catalog No. NC1832139
Encompass
Large Slide Warmer is ideal for use in cytology, histology, pathology, and biology for paraffin tissue section mounting. Anodized black surface provides contrast and even heat transfer. This Slide Warmer is ideal for use in cytology, histology, pathology, and biology for paraffin tissue section mounting. Anodized black surface provides contrast and even heat transfer. Thermostat setting is easy to use and can be set from room temperature to 75° C. Surface holds about 66 slides. Cord is CSA and UL approved. Surface measures 25” x 8” (63cm x 20cm). Unit weighs approximately 23 lbs.
Catalog No. NC1832139 Supplier Medicus Health Supplier No. 5311M2
Large Slide Warmer is ideal for use in cytology, histology, pathology, and biology for paraffin tissue section mounting. Anodized black surface provides contrast and even heat transfer. This Slide Warmer is ideal for use in cytology, histology, pathology, and biology for paraffin tissue section mounting. Anodized black surface provides contrast and even heat transfer. Thermostat setting is easy to use and can be set from room temperature to 75° C. Surface holds about 66 slides. Cord is CSA and UL approved. Surface measures 25” x 8” (63cm x 20cm). Unit weighs approximately 23 lbs.