Learn More
PATHOLOGY INNOVATIONS The 36 mm square stemless chuck is the same size as Pathology Innovations Large Chucks but made with a dovetail for cryostats that require stemless chucks.
Catalog No. NC3370402
Encompass_Preferred
These stainless steel chucks are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. Chucks can also be used warm with use of the “over-chuck freezing blocks”. These chucks were designed by professor and author of frozen section literature Dr. Stephen Peters to fit cryostats that require stemless chucks.
Catalog No. NC3370402 Supplier PATHOLOGY INNOVATIONS Supplier No. 151
These stainless steel chucks are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. Chucks can also be used warm with use of the “over-chuck freezing blocks”. These chucks were designed by professor and author of frozen section literature Dr. Stephen Peters to fit cryostats that require stemless chucks.