Learn More
Perkin Elmer US LLC Large Tin Capsule Kit, Pkg.250
Catalog No. 509052189
Encompass_Preferred
For Analysis of Nonvolatile Solid Samples Tin aids in the combustion of the sample. As it burns, tin releases heat, causing the sample to break down more quickly and more completely. Tin capsules are routinely used to analyze polymers, soils, coal, dry dairy products, plant leaves, paper, textiles, calibration standards, and many organic and pharmaceutical products. To use, simply place the sample in the tin capsule and fold the capsule closed. Choose standard size for most PerkinElmer 2400 applications and large size for most PerkinElmer 2410 applications.
Catalog No. 50-905-2189 Supplier Perkin Elmer US LLC Supplier No. N2411362
For Analysis of Nonvolatile Solid Samples Tin aids in the combustion of the sample. As it burns, tin releases heat, causing the sample to break down more quickly and more completely. Tin capsules are routinely used to analyze polymers, soils, coal, dry dairy products, plant leaves, paper, textiles, calibration standards, and many organic and pharmaceutical products. To use, simply place the sample in the tin capsule and fold the capsule closed. Choose standard size for most PerkinElmer 2400 applications and large size for most PerkinElmer 2410 applications.