Learn More
Ampco Large Valve Wheel Hook
SDP
Catalog No. 19175146 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
23 -5/8 in.
Catalog No. Length (English)
19-175-146 23 -5/8 in.
1 options
Catalog No. 19-175-146 Supplier Ampco Supplier No. WV40
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant valve wheel hook. Made in the USA.
  • F-type valve wheel hook
  • Aluminum bronze material, beryllium free.
  • Includes lifetime warranty.
  • Please see image for more information regarding measurements.
  • 23 and 5/8 in. length.
  • a (jaw opening size): 2 and 3/8 in.
  • b (throat depth): 4 in.
Specifications
Product Type Valve Wheel Hook
Length (English) 23 -5/8 in.
Quantity 1 Ea.