Learn More
McKesson General Medical LARGE VOLUME INFUSION PUMP INF
Catalog No. NC2250443
Encompass
Large Volume Infusion Pump Infusomat® Space® NiMH Rechargeable Battery Linear Peristaltic NonWireless 0.1 to 9999 mL Volume 0.1 to 1200 mL / Hr. Flow Rate
This item is not returnable. View return policy
Catalog No. NC2250443 Supplier McKesson General Medical Supplier No. 1034483
 Restricted Item
Add to Cart
Add to Cart
Large Volume Infusion Pump Infusomat® Space® NiMH Rechargeable Battery Linear Peristaltic NonWireless 0.1 to 9999 mL Volume 0.1 to 1200 mL / Hr. Flow Rate