Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Large Volume SPE Reservoir

Catalog No. 111009985 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PTFE
Polypropylene
Quantity:
3/Pk.
30/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Quantity
11-100-9985 PTFE 3/Pk.
11-101-0851 Polypropylene 30/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9985 Supplier MilliporeSigma™ Supelco™ Supplier No. 54259U
Add to Cart
Add to Cart

Large volume SPE reservoirs are designed to increase the head space volume of standard polypropylene SPE tubes. Because these reservoirs are designed to connect directly to the mouth of the SPE tube, they are ideal for gravity applicatioins where increased headspace volume is required.

TRUSTED_SUSTAINABILITY

Specifications

Description For use with 6 mL polypropylene SPE tubes
For Use With (Application) Food and Beverages
Type SPE Accessory
Volume (Metric) 25 mL
Material PTFE
Quantity 3/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.