Learn More
Perkin Elmer US LLC Large Volume Stainless Steel Capsule Kit
Supplier:  Perkin Elmer US LLC 03190021
Encompass_Preferred
Please note - these pans should not be heated above 300 °C. Any application requiring heating beyond 200 °C, or where gaseous decomposition products may be generated, should first be tested outside of the instrument. Pans contain the sample in a sealed environment throughout an experiment. The sealing suppresses the vaporization of a solvent or can contain a volatile reaction product, thereby eliminating the interfering effects of the heat of vaporization.
Catalog No. 50-211-2793